Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylpropanimidamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A197246-100G A197246-1G A197246-250MG A197246-25G A197246-5G
77274-674EA 286.01 USD InStock
77274-674 77274-676 77274-678 77274-680 77274-682
2-Methylpropanimidamide hydrochloride 95%
2-Methylpropanimidamide hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₁ClN₂
MW: 122.6 g/mol
MDL Number: MFCD00173787
CAS Number: 22007-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty