Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A197462-100G A197462-10G A197462-25G A197462-500G A197462-5G
77248-766EA 39 USD InStock
77248-766 77248-768 77248-770 77248-772 77746-244
3-Hydroxy-o-toluic acid 98%
3-Hydroxy-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00671541
CAS Number: 603-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty