Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluoropropionic acid 97%

Supplier: AMBEED, INC
Danger

A197546-100G A197546-10G A197546-1G A197546-25G A197546-5G
77248-780EA 475 USD InStock
77248-780 77248-782 77248-784 77248-786 77593-276
2,2-Difluoropropionic acid 97%
2,2-Difluoropropionic acid
Precaución: for Research Use Only


Formula: C₃H₄F₂O₂
MDL Number: MFCD03093764
CAS Number: 373-96-6
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty