Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A197566-10G A197566-25G A197566-500G A197566-5G A197566-100G
77237-078EA 16 USD InStock
77237-078 77237-080 77237-082 77237-084 77746-246
4-(Trifluoromethoxy)benzyl alcohol 97%
4-(Trifluoromethoxy)benzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₇F₃O₂
MW: 192.14
Density: 1.32
MDL Number: MFCD00036029
CAS Number: 1736-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty