Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino-PEG5-alcohol 97%

Supplier: AMBEED, INC
Warning

A199338-1G A199338-250MG A199338-25G
77260-746EA 87 USD InStock
77260-746 77260-748 77748-264
Amino-PEG5-alcohol 97%
Amino-PEG5-alcohol
Precaución: for Research Use Only


Formula: C₁₀H₂₃NO₅
MW: 237.293 g/mol
MDL Number: MFCD28950782
CAS Number: 34188-11-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty