Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cbz-4-Methylaminopieridine 97%

Supplier: AMBEED, INC
Warning

A200306-10G A200306-1G A200306-250MG A200306-5G
77287-644EA 90 USD InStock
77287-644 77287-646 77287-648 77287-650
1-Cbz-4-Methylaminopieridine 97%
1-Cbz-4-Methylaminopieridine
Precaución: for Research Use Only


Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD07772081
CAS Number: 405057-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty