Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl dithieno[3,2-b:2',3'-d]thiophene-2,6-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A200387-1G A200387-250MG A200387-5G
77237-110EA 46 USD InStock
77237-110 77237-112 77237-114
Diethyl dithieno[3,2-b:2',3'-d]thiophene-2,6-dicarboxylate 97%
Diethyl dithieno[3,2-b:2',3'-d]thiophene-2,6-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₄S₃
MDL Number: MFCD23103811
CAS Number: 502764-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty