Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethoxy-2-mercaptobenzimidazole 97%

Supplier: AMBEED, INC
Warning

A200623-10G A200623-25G A200623-5G
77248-840EA 35 USD InStock
77248-840 77248-842 77248-844
5-Ethoxy-2-mercaptobenzimidazole 97%
5-Ethoxy-2-mercaptobenzimidazole
Precaución: for Research Use Only


Formula: C₉H₁₀N₂OS
MDL Number: MFCD01326487
CAS Number: 55489-15-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty