Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-aminosalicylate dihydrate 98%

Supplier: AMBEED, INC
Warning

A200669-100G A200669-1MG A200669-25G
77287-666EA 42 USD InStock
77287-666 77287-668 77287-670
Sodium-4-aminosalicylate dihydrate 98%
Sodium 4-aminosalicylate dihydrate
Precaución: for Research Use Only


Formula: C₇H₆NNaO₃·2H₂O
MW: 211.14 g/mol (175.12 anhy)
MDL Number: MFCD00064392
CAS Number: 6018-19-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty