Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chlorotoluene 98%

Supplier: AMBEED, INC
Warning

A200741-100G A200741-1G A200741-500G A200741-5G A200741-25G
77287-672EA 48 USD InStock
77287-672 77287-674 77287-676 77287-678 77748-271
4-Bromo-2-chlorotoluene 98%
4-Bromo-2-chlorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrCl
MDL Number: MFCD00060651
CAS Number: 89794-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty