Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,3-Trifluoropropionic acid 97%

Supplier: AMBEED, INC
Danger

A200891-10G A200891-1G A200891-25G A200891-5G A200891-100G
77274-824EA 23 USD InStock
77274-824 77274-826 77274-828 77274-830 77748-272
3,3,3-Trifluoropropionic acid 97%
3,3,3-Trifluoropropionic acid
Precaución: for Research Use Only


Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 153
Melting Pt: 0
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty