Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4,5-Trifluorophenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A200914-100G A200914-10G A200914-25G A200914-500G
77237-142EA 17 USD InStock
77237-142 77237-144 77237-146 77237-148 77237-150
(2,4,5-Trifluorophenyl)acetic acid 98%
(2,4,5-Trifluorophenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₂F₃)CH₂COOH
MDL Number: MFCD00082479
CAS Number: 209995-38-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty