Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A202264-10G A202264-250MG A202264-25G A202264-5G A202264-1G
77274-886EA 43 USD InStock
77274-886 77274-888 77274-890 77274-892 77748-273
3-Chloro-5-fluorophenylboronic acid 97%
3-Chloro-5-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BClFO₂
MDL Number: MFCD06801741
CAS Number: 328956-61-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty