Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-Methylphenyl)sulfonamido)-N-(4-phenylthiazol-2-yl)-4-(trifluoromethyl)benzamide 98%

Supplier: AMBEED, INC
Warning

A202344-100MG A202344-10MG A202344-1MG A202344-25MG A202344-50MG A202344-5MG
77260-814EA 228 USD InStock
77260-814 77260-816 77260-818 77260-820 77260-822 77260-824
2-((4-Methylphenyl)sulfonamido)-N-(4-phenylthiazol-2-yl)-4-(trifluoromethyl)benzamide 98%
2-((4-Methylphenyl)sulfonamido)-N-(4-phenylthiazol-2-yl)-4-(trifluoromethyl)benzamide
Precaución: for Research Use Only


Formula: C₂₄H₁₈F₃N₃O₃S₂
MDL Number: MFCD30718174
CAS Number: 1991986-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty