Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-trifluoromethylbenzothiazole 98%

Supplier: AMBEED, INC
Warning

A202347-100MG A202347-1G A202347-250MG A202347-5G
77274-894EA 12 USD InStock
77274-894 77274-896 77274-898 77274-900
2-Chloro-6-trifluoromethylbenzothiazole 98%
2-Chloro-6-trifluoromethylbenzothiazole
Precaución: for Research Use Only


Formula: C₈H₃ClF₃NS
MW: 237.6293 g/mol
MDL Number: MFCD08459022
CAS Number: 159870-86-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty