Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydroxy-5-methoxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A205352-1G A205352-250MG A205352-25G A205352-5G
77248-930EA 20 USD InStock
77248-930 77248-932 77248-934 77248-936
3,4-Dihydroxy-5-methoxybenzaldehyde 95%
3,4-Dihydroxy-5-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₄
MDL Number: MFCD00016610
CAS Number: 3934-87-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty