Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-chloro-6-methylpyridine 97%

Supplier: AMBEED, INC
Warning

A205499-100MG A205499-1G A205499-250MG A205499-5G
77248-938EA 10 USD InStock
77248-938 77248-940 77248-942 77248-944
2-Amino-4-chloro-6-methylpyridine 97%
2-Amino-4-chloro-6-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂
MW: 142.5862 g/mol
MDL Number: MFCD00800671
CAS Number: 36340-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty