Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-ethoxypyridine 95%

Supplier: AMBEED, INC
Warning

A207390-10G A207390-1G A207390-250MG A207390-5G
77275-028EA 128 USD InStock
77275-028 77275-030 77275-032 77275-034
3-Bromo-2-ethoxypyridine 95%
3-Bromo-2-ethoxypyridine
Precaución: for Research Use Only


Formula: C₇H₈BrNO
MDL Number: MFCD00234309
CAS Number: 57883-25-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty