Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-nitrobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A208185-100G A208185-10G A208185-1G A208185-25G A208185-5G
77287-878EA 147 USD InStock
77287-878 77287-880 77287-882 77287-884 77287-886
4-Hydroxy-3-nitrobenzyl alcohol 98%
4-Hydroxy-3-nitrobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇NO₄
MW: 169.14
MDL Number: MFCD00024249
CAS Number: 41833-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty