Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(4-ethylphenyl)ethyne 98%

Supplier: AMBEED, INC
Warning

A208337-100MG A208337-1G A208337-250MG
77275-060EA 44 USD InStock
77275-060 77275-062 77275-064
1,2-Bis(4-ethylphenyl)ethyne 98%
1,2-Bis(4-ethylphenyl)ethyne
Precaución: for Research Use Only


Formula: C₁₈H₁₈
MDL Number: MFCD05864409
CAS Number: 79135-69-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty