Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ketotifen fumarate 95%

Supplier: AMBEED, INC
Warning

A208651-100MG A208651-1G A208651-250MG A208651-5G
77287-896EA 10 USD InStock
77287-896 77287-898 77287-900 77287-902 77287-904
Ketotifen fumarate 95%
Ketotifen fumarate
Precaución: for Research Use Only


Formula: C₁₉H₁₉NOS
MW: 425.50 g/mol
MDL Number: MFCD00079394
CAS Number: 34580-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty