Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N2-(1-Oxotetradecyl)-L-lysyl-L-alanyl-L-lysyl-L-alaninamide 97%

Supplier: AMBEED, INC
Warning

A208902-100MG A208902-250MG
77249-022EA 72 USD InStock
77249-022 77249-024
N2-(1-Oxotetradecyl)-L-lysyl-L-alanyl-L-lysyl-L-alaninamide 97%
N2-(1-Oxotetradecyl)-L-lysyl-L-alanyl-L-lysyl-L-alaninamide
Precaución: for Research Use Only


Formula: C₁₉H₁₇ClN₂O₄
CAS Number: 959610-24-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty