Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydroxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A209187-10G A209187-1G A209187-25G A209187-500G A209187-5G A209187-100G
77249-026EA 14 USD InStock
77249-026 77249-028 77249-030 77249-032 77249-034 77746-280
2,3-Dihydroxybenzaldehyde 97%
2,3-Dihydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₆O₃
MDL Number: MFCD00003324
CAS Number: 24677-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty