Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-bromo-3-chloro-5-fluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A210286-100MG A210286-250MG
77249-068EA 35 USD InStock
77249-068 77249-070
2-Amino-6-bromo-3-chloro-5-fluorobenzoic acid 97%
2-Amino-6-bromo-3-chloro-5-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO₂
MDL Number: MFCD23795936
CAS Number: 1513119-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty