Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-methylimidazo[1,2-b]pyridazine 97%

Supplier: AMBEED, INC
Danger

A210647-100MG A210647-1G A210647-250MG
77261-036EA 16 USD InStock
77261-036 77261-038 77261-040
6-Bromo-2-methylimidazo[1,2-b]pyridazine 97%
6-Bromo-2-methylimidazo[1,2-b]pyridazine
Precaución: for Research Use Only


Formula: C₇H₆BrN₃
MDL Number: MFCD29760607
CAS Number: 1936575-36-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty