Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dibromobenzo[d]thiazole 97%

Supplier: AMBEED, INC
Warning

A211640-100MG A211640-1G A211640-250MG
77261-074EA 36 USD InStock
77261-074 77261-076 77261-078
4,7-Dibromobenzo[d]thiazole 97%
4,7-Dibromobenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₇H₃Br₂NS
MDL Number: MFCD30528621
CAS Number: 2089292-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty