Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5aS,10bR)-2-(Perfluorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate 98%

Supplier: AMBEED, INC
Danger

A211721-100MG A211721-250MG
77249-122EA 34 USD InStock
77249-122 77249-124
(5aS,10bR)-2-(Perfluorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate 98%
(5aS,10bR)-2-(Perfluorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₈H₁₁BF₉N₃O
MDL Number: MFCD08459334
CAS Number: 740816-14-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty