Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(trans-4-Propylcyclohexyl)-4-iodobenzene 97%

Supplier: AMBEED, INC
Warning

A212912-10G A212912-25G A212912-5G
77275-226EA 29 USD InStock
77275-226 77275-228 77275-230 77275-232
1-(trans-4-Propylcyclohexyl)-4-iodobenzene 97%
1-(trans-4-Propylcyclohexyl)-4-iodobenzene
Precaución: for Research Use Only


Formula: C₁₅H₂₁I
MDL Number: MFCD06658188
CAS Number: 111158-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty