Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-methylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A213243-10G A213243-1G A213243-250MG A213243-25G A213243-5G
77249-152EA 65 USD InStock
77249-152 77249-154 77249-156 77249-158 77249-160
4-Hydroxy-3-methylbenzaldehyde 95%
4-Hydroxy-3-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₂
MW: 136.1479 g/mol
MDL Number: MFCD00012360
CAS Number: 15174-69-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty