Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-nitroquinoline 97%

Supplier: AMBEED, INC
Warning

A213480-1G A213480-25G A213480-5G
77261-138EA 8 USD InStock
77261-138 77261-140 77261-142
4-Chloro-3-nitroquinoline 97%
4-Chloro-3-nitroquinoline
Precaución: for Research Use Only


Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD07369246
CAS Number: 39061-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty