Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Chloro-3-(dimethylamino)allylidene)-N-methylmethanaminium hexafluorophosphate(V) 97%

Supplier: AMBEED, INC
Warning

A213654-100G A213654-10G A213654-25G A213654-500G A213654-5G
77261-144EA 38 USD InStock
77261-144 77261-146 77261-148 77261-150 77261-152
N-(2-Chloro-3-(dimethylamino)allylidene)-N-methylmethanaminium hexafluorophosphate(V) 97%
N-(2-Chloro-3-(dimethylamino)allylidene)-N-methylmethanaminium hexafluorophosphate(V)
Precaución: for Research Use Only


Formula: C₇H₁₄ClF₆N₂P
MDL Number: MFCD08458255
CAS Number: 249561-98-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty