Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-2'-(trifluoromethyl)acetanilide 97%

Supplier: AMBEED, INC
Warning

A214072-1G A214072-250MG
77261-156EA 29 USD InStock
77261-156 77261-158
2-Chloro-2'-(trifluoromethyl)acetanilide 97%
2-Chloro-2'-(trifluoromethyl)acetanilide
Precaución: for Research Use Only


Formula: C₉H₇ClF₃NO
MW: 237.61 g/mol
MDL Number: MFCD00018893
CAS Number: 3792-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty