Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-4-iodobenzene 98% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with copper chip

A214100-1000G A214100-100G A214100-10G A214100-25G A214100-500G A214100-5G
77288-056EA 233 USD InStock
77288-056 77288-058 77288-060 77288-062 77288-064 77288-066
1-Fluoro-4-iodobenzene 98% stabilised
1-Fluoro-4-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 182
Melting Pt: -18
MDL Number: MFCD00001052
CAS Number: 352-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty