Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A214129-1G A214129-250MG A214129-5G
77275-268EA 40 USD InStock
77275-268 77275-270 77275-272
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester 96%
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BO₅
MDL Number: MFCD06797992
CAS Number: 603122-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty