Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Baricitinib phosphate salt 98%

Supplier: AMBEED, INC
Warning

A214136-10MG A214136-1MG A214136-2MG A214136-5MG A214136-250MG
77237-478EA 30 USD InStock
77237-478 77237-480 77237-482 77237-484 77746-294
Baricitinib phosphate salt 98%
Baricitinib phosphate salt
Precaución: for Research Use Only


Formula: C₁₆H₁₇N₇O₂S·H₃PO₄
MDL Number: MFCD25976707
CAS Number: 1187595-84-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty