Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dipentyl carbonate 97%

Supplier: AMBEED, INC
Warning

A214148-100MG A214148-10G A214148-1G A214148-250MG A214148-25G A214148-5G
77237-486EA 17 USD InStock
77237-486 77237-488 77237-490 77237-492 77237-494 77237-496
Dipentyl carbonate 97%
Dipentyl carbonate
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₃
MDL Number: MFCD00068969
CAS Number: 2050-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty