Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-hydroxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A214333-1G A214333-250MG A214333-25G A214333-5G A214333-10G
77261-164EA 14 USD InStock
77261-164 77261-166 77261-168 77261-170 77748-335
2-Chloro-4-hydroxybenzonitrile 97%
2-Chloro-4-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₄ClNO
MDL Number: MFCD00052185
CAS Number: 3336-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty