Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A215154-100G A215154-10G A215154-25G A215154-5G
77275-274EA 41 USD InStock
77275-274 77275-276 77275-278 77275-280
2-Fluoro-4-methylpyridine 98%
2-Fluoro-4-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 157
MDL Number: MFCD00041224
CAS Number: 461-87-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty