Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((2,5-Dioxopyrrolidin-1-yl)oxy)-1,3-dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium hexafluorophosphate(V) 98%

Supplier: AMBEED, INC
Warning

A216375-1G A216375-25G A216375-5G
77249-240EA 10 USD InStock
77249-240 77249-242 77249-244
2-((2,5-Dioxopyrrolidin-1-yl)oxy)-1,3-dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium hexafluorophosphate(V) 98%
2-((2,5-Dioxopyrrolidin-1-yl)oxy)-1,3-dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium hexafluorophosphate(V)
Precaución: for Research Use Only


Formula: C₁₀H₁₆F₆N₃O₃P
MDL Number: MFCD08274619
CAS Number: 443305-33-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty