Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-m-toluic acid 97%

Supplier: AMBEED, INC
Warning

A216594-100G A216594-10G A216594-250MG A216594-25G A216594-5G
77275-322EA 95 USD InStock
77275-322 77275-324 77275-326 77275-328 77275-330
6-Fluoro-m-toluic acid 97%
6-Fluoro-m-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD00092819
CAS Number: 321-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty