Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KY1220 98%

Supplier: AMBEED, INC
Warning

A217096-10MG A217096-25MG A217096-2MG A217096-50MG A217096-5MG
77275-338EA 35 USD InStock
77275-338 77275-340 77275-342 77275-344 77275-346 77275-348
KY1220 98%
KY1220
Precaución: for Research Use Only


Formula: C₁₄H₁₀N₄O₃S
MDL Number: MFCD00609073
CAS Number: 292168-79-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty