Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OTS-964 98%

Supplier: AMBEED, INC
Warning

A217615-25MG A217615-50MG A217615-2MG
77237-580EA 77 USD InStock
77237-580 77237-582 77237-584 77746-300
OTS-964 98%
OTS-964
Precaución: for Research Use Only


Formula: C₂₂H₂₂N₂O₂S
MDL Number: MFCD28386220
CAS Number: 1338545-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty