Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Di-tert-butylbenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A217935-1G A217935-250MG A217935-25G A217935-5G
77275-388EA 26 USD InStock
77275-388 77275-390 77275-392 77275-394
3,5-Di-tert-butylbenzyl alcohol 98%
3,5-Di-tert-butylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₁₅H₂₄O
MDL Number: MFCD03844785
CAS Number: 77387-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty