Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A218418-1G A218418-250MG A218418-5G
77275-424EA 55 USD InStock
77275-424 77275-426 77275-428
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid 98%
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₅BrN₂O₂
MW: 241.05 g/mol (anhy)
MDL Number: MFCD07021511
CAS Number: 886362-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty