Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trifluorobenzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A220276-10G A220276-1G A220276-250MG
77275-458EA 42 USD InStock
77275-458 77275-460 77275-462
3,4,5-Trifluorobenzenesulphonyl chloride 98%
3,4,5-Trifluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₂ClF₃O₂S
MW: 230.59
MDL Number: MFCD03094559
CAS Number: 351003-43-5
UN: 2924
ADR: 3,8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty