Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromo-4-fluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A220368-10G A220368-1G A220368-25G A220368-100G A220368-5G
77275-466EA 17 USD InStock
77275-466 77275-468 77275-470 77748-352 77748-353
Methyl-3-bromo-4-fluorobenzoate 97%
Methyl-3-bromo-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD04972915
CAS Number: 82702-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty