Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-4,4-difluoroheptanedioate 97%

Supplier: AMBEED, INC
Warning

A221206-10G A221206-1G A221206-250MG A221206-5G A221206-25G
77288-226EA 157 USD InStock
77288-226 77288-228 77288-230 77288-232 77748-356
Diethyl-4,4-difluoroheptanedioate 97%
Diethyl-4,4-difluoroheptanedioate
Precaución: for Research Use Only


Formula: C₁₁H₁₈F₂O₄
MW: 252.26 g/mol
MDL Number: MFCD08146634
CAS Number: 22515-16-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty