Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-5-(trifluoromethyl)picolinic acid 97%

Supplier: AMBEED, INC
Warning

A222308-100MG A222308-1G A222308-250MG A222308-5G A222308-50MG
77261-312EA 149 USD InStock
77261-312 77261-314 77261-316 77261-318 77748-359
3-Hydroxy-5-(trifluoromethyl)picolinic acid 97%
3-Hydroxy-5-(trifluoromethyl)picolinic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₃
MW: 207.10 g/mol
MDL Number: MFCD28368540
CAS Number: 1806579-86-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty