Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Nitrophenyl)propionamide 97%

Supplier: AMBEED, INC
Warning

A222636-10G A222636-1G A222636-250MG
77261-326EA 34 USD InStock
77261-326 77261-328 77261-330
N-(3-Nitrophenyl)propionamide 97%
N-(3-Nitrophenyl)propionamide
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O₃
MDL Number: MFCD00791259
CAS Number: 7470-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty