Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Furylmethylamine 98%

Supplier: AMBEED, INC
Danger

A223239-100MG A223239-1G A223239-250MG
77275-534EA 49 USD InStock
77275-534 77275-536 77275-538
3-Furylmethylamine 98%
3-Furylmethylamine
Precaución: for Research Use Only


Formula: C₅H₇NO
MDL Number: MFCD02681983
CAS Number: 4543-47-9
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty